Ethyl 2-formylthiazole-4-carboxylate, tech

Lieferant: Apollo Scientific
Achtung

OR322913-250MG OR322913-1G
APOSOR322913-250MGEA 48 EUR
APOSOR322913-250MG APOSOR322913-1G
Ethyl 2-formylthiazole-4-carboxylate, tech
Ethyl 2-formylthiazole-4-carboxylate
Formel: C₇H₇NO₃S
Molecular Weight: 185.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689563
CAS-Nummer: 73956-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR