8-Methylquinazoline-2,4(1h,3h)-dione 97%

Lieferant: Apollo Scientific

Synonyme: 8-Methylquinazoline-2,4(1H,3H)-dione

OR323138-5G
APOSOR323138-5GEA 161 EUR
APOSOR323138-5G
8-Methylquinazoline-2,4(1h,3h)-dione 97%
8-Methylquinazoline-2,4(1h,3h)-dione
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02167411
CAS-Nummer: 67449-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR