4-Methyl-2-chinolinol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Lepidinol, 4-Methylquinolin-2-ol, 2-Hydroxy-4-methylquinoline

OR32653-100G OR32653-25G OR32653-5G
APOSOR32653-100GEA 213 EUR
APOSOR32653-100G APOSOR32653-25G APOSOR32653-5G
4-Methyl-2-chinolinol 95%
4-Methyl-2-chinolinol
Formel: C₁₀H₉NO
Molecular Weight: 159,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006745
CAS-Nummer: 84909-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR