(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one 95%
Lieferant: Apollo Scientific
Synonyme:
(E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one
OR32678-1G
OR32678-5G
APOSOR32678-1GEA
336
EUR
APOSOR32678-1G
APOSOR32678-5G
(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one 95%
(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one
Formel:
C₉H₁₀BrNOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD03618050 CAS-Nummer: 1422045-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...