3-(1H-Pyrrol-1-yl)-1-benzofuran-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR32685-1G OR32685-5G
APOSOR32685-1GEA 538 EUR
APOSOR32685-1G APOSOR32685-5G
3-(1H-Pyrrol-1-yl)-1-benzofuran-2-carboxylic acid 95%
3-(1H-Pyrrol-1-yl)-1-benzofuran-2-carboxylic acid
Formel: C₁₃H₉NO₃
Molecular Weight: 227.215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00445255
CAS-Nummer: 80066-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR