5-Bromoquinoline-8-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-8-formyl-1-azanaphthalene, 5-Bromoquinoline-8-carbaldehyde, 5-Bromo-8-formylquinoline

OR3274-1G OR3274-5G
APOSOR3274-1GEA 46.4 EUR
APOSOR3274-1G APOSOR3274-5G
5-Bromoquinoline-8-carboxaldehyde
5-Bromoquinoline-8-carboxaldehyde
Formel: C₁₀H₆BrNO
Molecular Weight: 236.065 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08064042
CAS-Nummer: 885267-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR