Ethyl 4-hydroxy-6-methylquinoline-3-carboxylate 95%

Lieferant: Apollo Scientific

OR32853-1G OR32853-5G
APOSOR32853-1GEA 114 EUR
APOSOR32853-1G APOSOR32853-5G
Ethyl 4-hydroxy-6-methylquinoline-3-carboxylate 95%
Ethyl 4-hydroxy-6-methylquinoline-3-carboxylate
Formel: C₁₃H₁₃NO₃
Molecular Weight: 231,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00087702
CAS-Nummer: 85418-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR