Ethyl 2-(2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)acetate 95%

Lieferant: Apollo Scientific

OR32868-1G OR32868-5G
APOSOR32868-1GEA 339 EUR
APOSOR32868-1G APOSOR32868-5G
Ethyl 2-(2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)acetate 95%
Ethyl 2-(2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)acetate
Formel: C₁₁H₁₁NO₄
Molecular Weight: 221.209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00823452
CAS-Nummer: 13610-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR