1H-Indole-3-carboxamide 95%

Lieferant: Apollo Scientific

OR32891-1G OR32891-5G
APOSOR32891-1GEA 163 EUR
APOSOR32891-1G APOSOR32891-5G
1H-Indole-3-carboxamide 95%
1H-Indole-3-carboxamide
Formel: C₉H₈N₂O
Molecular Weight: 160,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00460642
CAS-Nummer: 1670-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR