2-(Chloromethyl)-5-hydroxy-4H-pyran-4-one 95%

Lieferant: Apollo Scientific

OR32940-1G OR32940-5G
APOSOR32940-1GEA 339 EUR
APOSOR32940-1G APOSOR32940-5G
2-(Chloromethyl)-5-hydroxy-4H-pyran-4-one 95%
2-(Chloromethyl)-5-hydroxy-4H-pyran-4-one
Formel: C₆H₅ClO₃
Molecular Weight: 160,555 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053592
CAS-Nummer: 7559-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR