3-Methyl-3,4-dihydro-1,2,3-benzotriazin-4-one 95%

Lieferant: Apollo Scientific

OR32990-1G
APOSOR32990-1GEA 339 EUR
APOSOR32990-1G
3-Methyl-3,4-dihydro-1,2,3-benzotriazin-4-one 95%
3-Methyl-3,4-dihydro-1,2,3-benzotriazin-4-one
Formel: C₈H₇N₃O
Molecular Weight: 161,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125779
CAS-Nummer: 22305-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR