3-Bromo-5-carboxybenzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Borono-5-bromobenzoic acid, 3-Bromo-5-carboxyphenylboronic acid

OR3302-25G OR3302-5G
APOSOR3302-25GEA 261 EUR
APOSOR3302-25G APOSOR3302-5G
3-Bromo-5-carboxybenzeneboronic acid 97%
3-Bromo-5-carboxybenzeneboronic acid
Formel: C₇H₆BBrO₄
Molecular Weight: 244,835 g/mol
Schmelzpunkt: 290…292 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689487
CAS-Nummer: 913835-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR