[(2-Chlorophenyl)methyl]urea 95%

Lieferant: Apollo Scientific

OR33122-1G
APOSOR33122-1GEA 339 EUR
APOSOR33122-1G
[(2-Chlorophenyl)methyl]urea 95%
[(2-Chlorophenyl)methyl]urea
Formel: C₈H₉ClN₂O
Molecular Weight: 184,623 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01314628
CAS-Nummer: 4654-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR