5-Bromo-3-chloro-2-hydrazinopyridine

Lieferant: Apollo Scientific
Achtung

OR33680-1G OR33680-250MG OR33680-5G
APOSOR33680-1GEA 348 EUR
APOSOR33680-1G APOSOR33680-250MG APOSOR33680-5G
5-Bromo-3-chloro-2-hydrazinopyridine
5-Bromo-3-chloro-2-hydrazinopyridine
Formel: C₅H₅BrClN₃
Molecular Weight: 222.47 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10000107
CAS-Nummer: 1289076-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR