3-Hydrazinobenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Cyanophenylhydrazine
OR3388-5G
OR3388-100G
OR3388-25G
APOSOR3388-5GEA
33.5
EUR
APOSOR3388-5G
APOSOR3388-100G
APOSOR3388-25G
3-Hydrazinobenzonitrile
3-Hydrazinobenzonitrile
Formel:
C₇H₇N₃ Molecular Weight: 133,151 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03030048 CAS-Nummer: 17672-26-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...