Thieno[3,2-b]pyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR34493-1G OR34493-250MG
APOSOR34493-1GEA 208 EUR
APOSOR34493-1G APOSOR34493-250MG
Thieno[3,2-b]pyridine-2-carbohydrazide
Thieno[3,2-b]pyridine-2-carbohydrazide
Formel: C₈H₇N₃OS
Molecular Weight: 193,226 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32066838
CAS-Nummer: 2391987-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR