2,6-Dichloro-4-methoxypyridine 99%

Lieferant: Apollo Scientific
Achtung

OR345036-1G OR345036-5G
APOSOR345036-1GEA 23.2 EUR
APOSOR345036-1G APOSOR345036-5G
2,6-Dichloro-4-methoxypyridine 99%
2,6-Dichloro-4-methoxypyridine
Formel: C₆H₅Cl₂NO
Molecular Weight: 178.016 g/mol
Schmelzpunkt: 108…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00955755
CAS-Nummer: 17228-75-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR