4-(2-Hydroxyethyl)benzoic acid 95%

Lieferant: Apollo Scientific

OR345096-1G OR345096-5G
APOSOR345096-1GEA 72.5 EUR
APOSOR345096-1G APOSOR345096-5G
4-(2-Hydroxyethyl)benzoic acid 95%
4-(2-Hydroxyethyl)benzoic acid
Formel: C₉H₁₀O₃
Molecular Weight: 166.174 g/mol
Schmelzpunkt: 122…129 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02114379
CAS-Nummer: 46112-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR