3,5-Dibromo-2-nitropyridine

Lieferant: Apollo Scientific
Achtung

OR345119-1G OR345119-250MG
APOSOR345119-1GEA 214 EUR
APOSOR345119-1G APOSOR345119-250MG
3,5-Dibromo-2-nitropyridine
3,5-Dibromo-2-nitropyridine
Formel: C₅H₂Br₂N₂O₂
Molecular Weight: 281,89 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01612785
CAS-Nummer: 610261-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR