2-Bromo-5-chloro-3-hydroxypyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-chloropyridin-3-ol
OR345241-1G
OR345241-250MG
OR345241-5G
APOSOR345241-1GEA
136
EUR
APOSOR345241-1G
APOSOR345241-250MG
APOSOR345241-5G
2-Bromo-5-chloro-3-hydroxypyridine 95%
2-Bromo-5-chloro-3-hydroxypyridine
Formel:
C₅H₃BrClNO Molecular Weight: 208.44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835245 CAS-Nummer: 127561-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...