2-Methyl-3-nitropyridine-6-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR345306-250MG OR345306-1G
APOSOR345306-250MGEA 139 EUR
APOSOR345306-250MG APOSOR345306-1G
2-Methyl-3-nitropyridine-6-carboxylic acid 95%
2-Methyl-3-nitropyridine-6-carboxylic acid
Formel: C₇H₆N₂O₄
Molecular Weight: 182.134 g/mol
Schmelzpunkt: 123…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07776980
CAS-Nummer: 24194-98-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR