4-Chloro-2-methyl-7-azaindole 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine

OR345316-1G OR345316-250MG OR345316-10G
APOSOR345316-1GEA 98.6 EUR
APOSOR345316-1G APOSOR345316-250MG APOSOR345316-10G
4-Chloro-2-methyl-7-azaindole 95%
4-Chloro-2-methyl-7-azaindole
Formel: C₈H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12924600
CAS-Nummer: 307951-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR