3-Hydroxy-5-methylbenzoic acid 95%

Lieferant: Apollo Scientific

OR345377-25G OR345377-5G
APOSOR345377-25GEA 439 EUR
APOSOR345377-25G APOSOR345377-5G
3-Hydroxy-5-methylbenzoic acid 95%
3-Hydroxy-5-methylbenzoic acid
Formel: C₈H₈O₃
Molecular Weight: 152,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09833213
CAS-Nummer: 585-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR