4-{[(Morpholin-4-yl)carbonyl]amino}benzeneboronic acid, pinacol ester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamoyl}morpholine, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine-4-carboxamide
OR3454-1G
OR3454-5G
APOSOR3454-1GEA
191
EUR
APOSOR3454-1G
APOSOR3454-5G
4-{[(Morpholin-4-yl)carbonyl]amino}benzeneboronic acid, pinacol ester 98%
4-{[(Morpholin-4-yl)carbonyl]amino}benzeneboronic acid, pinacol ester
Formel:
C₁₇H₂₅BN₂O₄ Molecular Weight: 332,202 g/mol Schmelzpunkt: 248…250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689495 CAS-Nummer: 874290-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...