4-Hydroxy-2-nitrobenzoic acid 95%
Lieferant: Apollo Scientific
OR345405-1G
OR345405-250MG
APOSOR345405-1GEA
0
EUR
APOSOR345405-1G
APOSOR345405-250MG
4-Hydroxy-2-nitrobenzoic acid 95%
4-Hydroxy-2-nitrobenzoic acid
Formel:
C₇H₅NO₅ Schmelzpunkt: 230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09908185 CAS-Nummer: 74230-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...