5-Bromo-4-methoxy-2-methylbenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR345438-5G
OR345438-1G
APOSOR345438-5GEA
683
EUR
APOSOR345438-5G
APOSOR345438-1G
5-Bromo-4-methoxy-2-methylbenzoic acid 95%
5-Bromo-4-methoxy-2-methylbenzoic acid
Formel:
C₉H₉BrO₃ Molecular Weight: 245.07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044844 CAS-Nummer: 875245-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...