3-Bromo-2-cyano-5-methoxypyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-5-methoxypicolinonitrile
OR345524-1G
OR345524-250MG
APOSOR345524-1GEA
261
EUR
APOSOR345524-1G
APOSOR345524-250MG
3-Bromo-2-cyano-5-methoxypyridine 97%
3-Bromo-2-cyano-5-methoxypyridine
Formel:
C₇H₅BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062679 CAS-Nummer: 717843-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...