6-Bromo-3,4-methylenedioxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-1,3-benzodioxole-5-carboxylic acid, 6-Bromobenzo[d][1,3]dioxole-5-carboxylic acid

OR345600-1G OR345600-25G OR345600-5G
APOSOR345600-1GEA 72.5 EUR
APOSOR345600-1G APOSOR345600-25G APOSOR345600-5G
6-Bromo-3,4-methylenedioxybenzoic acid
6-Bromo-3,4-methylenedioxybenzoic acid
Formel: C₈H₅BrO₄
Molecular Weight: 245,027 g/mol
Schmelzpunkt: 201…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156994
CAS-Nummer: 60546-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR