4,5-Dichloro-2,6-dimethylpyrimidine

Lieferant: Apollo Scientific

OR345711-1G OR345711-250MG
APOSOR345711-1GEA 190 EUR
APOSOR345711-1G APOSOR345711-250MG
4,5-Dichloro-2,6-dimethylpyrimidine
4,5-Dichloro-2,6-dimethylpyrimidine
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177.031 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16171717
CAS-Nummer: 105742-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR