3-(2,6-Dichloro-phenyl)-5-methyl-isoxazole-4-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR346020-1G OR346020-5G
APOSOR346020-1GEA 0 EUR
APOSOR346020-1G APOSOR346020-5G
3-(2,6-Dichloro-phenyl)-5-methyl-isoxazole-4-carboxylic acid ethyl ester
3-(2,6-Dichloro-phenyl)-5-methyl-isoxazole-4-carboxylic acid ethyl ester
Formel: C₁₃H₁₁Cl₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00115543
CAS-Nummer: 24248-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR