Ethyl 5-thiophen-2-yl-[1,2,4]oxadiazole-3-carboxylate
Lieferant: Apollo Scientific
OR346025-1G
OR346025-5G
APOSOR346025-1GEA
0
EUR
APOSOR346025-1G
APOSOR346025-5G
Ethyl 5-thiophen-2-yl-[1,2,4]oxadiazole-3-carboxylate
Ethyl 5-thiophen-2-yl-[1,2,4]oxadiazole-3-carboxylate
Formel:
C₉H₈N₂O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD10568211 CAS-Nummer: 40019-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...