1-tert-Butyloxycarbonyl-3-(N-Hydroxycarbamimidoyl)piperidine
Lieferant: Apollo Scientific
OR346055-1G
OR346055-500MG
OR346055-5G
APOSOR346055-1GEA
0
EUR
APOSOR346055-1G
APOSOR346055-500MG
APOSOR346055-5G
1-tert-Butyloxycarbonyl-3-(N-Hydroxycarbamimidoyl)piperidine
1-tert-Butyloxycarbonyl-3-(N-Hydroxycarbamimidoyl)piperidine
Formel:
C₁₁H₂₁N₃O₃ Molecular Weight: 243,303 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09951952 CAS-Nummer: 479080-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...