N'-Hydroxy-2-methoxy-benzamidine
Lieferant: Apollo Scientific
OR346058-1G
OR346058-5G
APOSOR346058-1GEA
0
EUR
APOSOR346058-1G
APOSOR346058-5G
N'-Hydroxy-2-methoxy-benzamidine
N'-Hydroxy-2-methoxy-benzamidine
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166.177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06366748 CAS-Nummer: 771-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...