Ethyl 5-(1-hydroxyethyl)-3-phenylisoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR346089-1G OR346089-5G
APOSOR346089-1GEA 0 EUR
APOSOR346089-1G APOSOR346089-5G
Ethyl 5-(1-hydroxyethyl)-3-phenylisoxazole-4-carboxylate
Ethyl 5-(1-hydroxyethyl)-3-phenylisoxazole-4-carboxylate
Formel: C₁₄H₁₅NO₄
Molecular Weight: 261,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00833029
CAS-Nummer: 95104-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR