Acetic acid 1,2-dibromo-ethyl ester

Lieferant: Apollo Scientific

OR346136-1G OR346136-5G
APOSOR346136-1GEA 0 EUR
APOSOR346136-1G APOSOR346136-5G
Acetic acid 1,2-dibromo-ethyl ester
Acetic acid 1,2-dibromo-ethyl ester
Formel: C₄H₆Br₂O₂
Molecular Weight: 245,897 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457744
CAS-Nummer: 24442-57-7
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR