6-Bromo-3-hydroxy-benzo[b]thiophene-2-carboxylic acid methyl ester

Lieferant: Apollo Scientific

OR346156-1G OR346156-5G OR346156-10G
APOSOR346156-1GEA 0 EUR
APOSOR346156-1G APOSOR346156-5G APOSOR346156-10G
6-Bromo-3-hydroxy-benzo[b]thiophene-2-carboxylic acid methyl ester
6-Bromo-3-hydroxy-benzo[b]thiophene-2-carboxylic acid methyl ester
Formel: C₁₀H₇BrO₃S
Molecular Weight: 287.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845695
CAS-Nummer: 863904-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR