3-Hydroxy-5-methylsulphanyl-isothiazole-4-carboxylic acid methyl ester
Lieferant: Apollo Scientific
OR346177-1G OR346177-500MG
APOSOR346177-1GEA 0 EUR
APOSOR346177-1G APOSOR346177-500MG
3-Hydroxy-5-methylsulphanyl-isothiazole-4-carboxylic acid methyl ester
3-Hydroxy-5-methylsulphanyl-isothiazole-4-carboxylic acid methyl ester
Formel: C₆H₇NO₃S₂
Molecular Weight: 205.255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501431
CAS-Nummer: 878477-22-6