5-tert-Butyl-[1,2,4]oxadiazole-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 5-(tert-butyl)-1,2,4-oxadiazole-3-carboxylate

OR346202-1G OR346202-500MG
APOSOR346202-1GEA 0 EUR
APOSOR346202-1G APOSOR346202-500MG
5-tert-Butyl-[1,2,4]oxadiazole-3-carboxylic acid ethyl ester
5-tert-Butyl-[1,2,4]oxadiazole-3-carboxylic acid ethyl ester
Formel: C₉H₁₄N₂O₃
Molecular Weight: 198.219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11975623
CAS-Nummer: 158154-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR