1-Phenyl-1H-pyrrole-2-carbaldehyde

Lieferant: Apollo Scientific

OR346239-1G OR346239-5G
APOSOR346239-1GEA 0 EUR
APOSOR346239-1G APOSOR346239-5G
1-Phenyl-1H-pyrrole-2-carbaldehyde
1-Phenyl-1H-pyrrole-2-carbaldehyde
Formel: C₁₁H₉NO
Molecular Weight: 171.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00457595
CAS-Nummer: 30186-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR