Benzofuran-3-carboxylic acid methyl ester

Lieferant: Apollo Scientific
Achtung

OR346248-1G OR346248-500MG OR346248-5G
APOSOR346248-1GEA 0 EUR
APOSOR346248-1G APOSOR346248-500MG APOSOR346248-5G
Benzofuran-3-carboxylic acid methyl ester
Benzofuran-3-carboxylic acid methyl ester
Formel: C₁₀H₈O₃
Molecular Weight: 176.169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15144245
CAS-Nummer: 4687-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR