(E)-3-Bromo-2-methyl-acrylic acid methyl ester

Lieferant: Apollo Scientific

OR346254-1G OR346254-5G
APOSOR346254-1GEA 0 EUR
APOSOR346254-1G APOSOR346254-5G
(E)-3-Bromo-2-methyl-acrylic acid methyl ester
(E)-3-Bromo-2-methyl-acrylic acid methyl ester
Formel: C₅H₇BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00154501
CAS-Nummer: 40053-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR