1-(2-Chloro-4-methylsulphanyl-phenyl)-ethanone
Lieferant: Apollo Scientific
OR346257-1G
OR346257-5G
APOSOR346257-1GEA
0
EUR
APOSOR346257-1G
APOSOR346257-5G
1-(2-Chloro-4-methylsulphanyl-phenyl)-ethanone
1-(2-Chloro-4-methylsulphanyl-phenyl)-ethanone
Formel:
C₉H₉ClOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD19981474 CAS-Nummer: 35515-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...