2,4-Dioxo-4-phenyl-butyric acid ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2,4-dioxo-4-phenylbutanoate

OR346270-1G OR346270-5G
APOSOR346270-1GEA 56.7 EUR
APOSOR346270-1G APOSOR346270-5G
2,4-Dioxo-4-phenyl-butyric acid ethyl ester
2,4-Dioxo-4-phenyl-butyric acid ethyl ester
Formel: C₁₂H₁₂O₄
Molecular Weight: 220.221 g/mol
Schmelzpunkt: 36…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01935977
CAS-Nummer: 6296-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR