4-Bromo-3-methyl-isothiazol-5-ylamine

Lieferant: Apollo Scientific

OR346313-1G OR346313-5G
APOSOR346313-1GEA 0 EUR
APOSOR346313-1G APOSOR346313-5G
4-Bromo-3-methyl-isothiazol-5-ylamine
4-Bromo-3-methyl-isothiazol-5-ylamine
Formel: C₄H₅BrN₂S
Molecular Weight: 193,065 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159755
CAS-Nummer: 85508-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR