3-Methyl-isoxazole-5-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR346323-1G OR346323-5G
APOSOR346323-1GEA 0 EUR
APOSOR346323-1G APOSOR346323-5G
3-Methyl-isoxazole-5-carboxylic acid ethyl ester
3-Methyl-isoxazole-5-carboxylic acid ethyl ester
Formel: C₇H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11656615
CAS-Nummer: 63366-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR