3-Methyl-isothiazol-4-ylamine

Lieferant: Apollo Scientific
Achtung

OR346368-1G OR346368-250MG OR346368-50MG
APOSOR346368-1GEA 621 EUR
APOSOR346368-1G APOSOR346368-250MG APOSOR346368-50MG
3-Methyl-isothiazol-4-ylamine
3-Methyl-isothiazol-4-ylamine
Formel: C₄H₆N₂S
Molecular Weight: 114,169 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10700204
CAS-Nummer: 53483-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR