5-Phenyl-1H-pyrazolo[3,4-c]pyridazin-3-ylamine

Lieferant: Apollo Scientific

Synonyme: 5-Phenyl-1H-pyrazolo[3,4-c]pyridazin-3-amine

OR346373-250MG OR346373-1G
APOSOR346373-250MGEA 0 EUR
APOSOR346373-250MG APOSOR346373-1G
5-Phenyl-1H-pyrazolo[3,4-c]pyridazin-3-ylamine
5-Phenyl-1H-pyrazolo[3,4-c]pyridazin-3-ylamine
Formel: C₁₁H₉N₅
Molecular Weight: 211,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23115309
CAS-Nummer: 405224-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR