2-tert-Butyl-5-chloro-6-methyl-pyrimidin-4-ol 95+%

Lieferant: Apollo Scientific

OR346383-1G OR346383-5G
APOSOR346383-1GEA 142 EUR
APOSOR346383-1G APOSOR346383-5G
2-tert-Butyl-5-chloro-6-methyl-pyrimidin-4-ol 95+%
2-tert-Butyl-5-chloro-6-methyl-pyrimidin-4-ol
Formel: C₉H₁₃ClN₂O
Molecular Weight: 200.665 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00114448
CAS-Nummer: 287927-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR