2-Chloro-4-methylnicotinic acid ethyl ester 95+%
Lieferant: Apollo Scientific
Synonyme: Ethyl 2-chloro-4-methylnicotinate
OR346414-1G OR346414-500MG
APOSOR346414-1GEA 497 EUR
APOSOR346414-1G APOSOR346414-500MG
2-Chloro-4-methylnicotinic acid ethyl ester 95+%
2-Chloro-4-methylnicotinic acid ethyl ester
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501333
CAS-Nummer: 50840-02-3