2-Amino-4,5-dimethyl-1-phenyl-1H-pyrrole-3-carbonitrile 95+%

Lieferant: Apollo Scientific

OR346420-1G OR346420-500MG OR346420-5G
APOSOR346420-1GEA 318 EUR
APOSOR346420-1G APOSOR346420-500MG APOSOR346420-5G
2-Amino-4,5-dimethyl-1-phenyl-1H-pyrrole-3-carbonitrile 95+%
2-Amino-4,5-dimethyl-1-phenyl-1H-pyrrole-3-carbonitrile
Formel: C₁₃H₁₃N₃
Molecular Weight: 211,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08691116
CAS-Nummer: 54329-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR